Learn More
MSC Rubbermaid Recycling Container, BRUTE, 32 gal, Round, Green, 1 ea
Catalog No. 502866443
Encompass
This vented BRUTE container is designed for efficient waste management. Its durable construction includes UV inhibitors, ensuring longevity without fading, warping, cracking, or crushing. Innovative features enhance productivity and user safety.
  • Venting channels reduce effort for liner removal by up to 50%
  • Integrated cinches secure liners for efficient, knot-free changes
  • Rounded and reinforced handles allow for easier lifting of heavy loads
  • Reinforced base engineered for dragging over rough surfaces
  • Contoured base handles improve grip and ergonomics
  • Integrated rib structure on rim increases strength and resists crushing
  • Stackable design for easy transport and storage
Catalog No. 50-286-6443 Supplier MSC Supplier No. 61633194
This vented BRUTE container is designed for efficient waste management. Its durable construction includes UV inhibitors, ensuring longevity without fading, warping, cracking, or crushing. Innovative features enhance productivity and user safety.
  • Venting channels reduce effort for liner removal by up to 50%
  • Integrated cinches secure liners for efficient, knot-free changes
  • Rounded and reinforced handles allow for easier lifting of heavy loads
  • Reinforced base engineered for dragging over rough surfaces
  • Contoured base handles improve grip and ergonomics
  • Integrated rib structure on rim increases strength and resists crushing
  • Stackable design for easy transport and storage