Learn More
Biotium 5-Bromouridine-5'-Triphosphate, 10 mM in TE Buffer
Catalog No. 501964634
Encompass_Preferred
BrUTP can be enzymatically incorporated into RNA. It is reported to be an excellent substrate for RNA polymerase and has been used to monitor nucleolar transcription in situ. Properties: Clear colorless liquid; Store at -20⁰C, protect from light; C9H11BrN2Na3O15P3; MW: 628.98; [161848-60-8].
Catalog No. 50-196-4634 Supplier Biotium Supplier No. 40026
BrUTP can be enzymatically incorporated into RNA. It is reported to be an excellent substrate for RNA polymerase and has been used to monitor nucleolar transcription in situ. Properties: Clear colorless liquid; Store at -20⁰C, protect from light; C9H11BrN2Na3O15P3; MW: 628.98; [161848-60-8].