Learn More
Aviva Systems Biology Corp BRWD1 Antibody - N-terminal region (ARP41298_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41298_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against BRWD1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human BRWD1. Alternate names: C21orf107, FLJ43918, N143, WDR9ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-57798