Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Bryodulcosigenin | 88930-16-9 | 1 MG
SDP

Catalog No. 5000421433
Encompass_Preferred

Bryodulcosigenin is an extract from the roots of Bryonia dioica with an anti-inflammatory effect. It is a solid, white to off-white compound with 99.21% purity and a molecular weight of 474.72. It is classified as a terpenoid triterpene.

  • Purity: 99.21%
  • Molecular weight: 474.72
  • Formula: C30H50O4
  • Soluble in DMSO (100 mg/mL, requires ultrasonic)
  • Can be prepared for in vivo dissolution in 10% DMSO and 90% (20% SBE-β-CD in Saline) or 10% DMSO and 90% Corn Oil.

Catalog No. 50-004-21433 Supplier Medchemexpress LLC Supplier No. HYN43121MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bryodulcosigenin is an extract from the roots of Bryonia dioica with an anti-inflammatory effect. It is a solid, white to off-white compound with 99.21% purity and a molecular weight of 474.72. It is classified as a terpenoid triterpene.

  • Purity: 99.21%
  • Molecular weight: 474.72
  • Formula: C30H50O4
  • Soluble in DMSO (100 mg/mL, requires ultrasonic)
  • Can be prepared for in vivo dissolution in 10% DMSO and 90% (20% SBE-β-CD in Saline) or 10% DMSO and 90% Corn Oil.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.