Learn More
Enzo Life Sciences Bryostatin 1 (10 µg). CAS: 83314-01-6
SDP
Supplier:  Enzo Life Sciences BMLST1030010
Encompass_Preferred
A structurally unique marine natural product that binds to and activates PKC at picomolar concentrations, but is not a carcinogen or a complete tumor promoter. In many systems it induces only a subset of the responses to PMA and blocks those which it does not induce. It is markedly more potent than PMA for translocating PKCδ and -ε. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2151