Learn More
Being Scientific BS-1 - 10.0 x 10.0 inch, 40~250rpm
Catalog No. 5000217592
Encompass_Preferred
BEING BS Series Orbital Shakers offer 3 sizes to fit your capacity requirements. Accuracy in speed and run time are guaranteed by the PID microprocessor controller. Programmability is a feature that will allow a variety of shaking protocols to be used. A full line of accessories is available to accommodate a variety of tubes, flasks, and other containers. As always BEING makes sure that all our shakers come with the necessary features and alarms guarantying your safety when in operation. Robust in design, construction, and mechanics, the BS series shakers will provide many years of service for your shaking needs.
Catalog No. 50-002-17592 Supplier Being Scientific Supplier No. OS1511U
BEING BS Series Orbital Shakers offer 3 sizes to fit your capacity requirements. Accuracy in speed and run time are guaranteed by the PID microprocessor controller. Programmability is a feature that will allow a variety of shaking protocols to be used. A full line of accessories is available to accommodate a variety of tubes, flasks, and other containers. As always BEING makes sure that all our shakers come with the necessary features and alarms guarantying your safety when in operation. Robust in design, construction, and mechanics, the BS series shakers will provide many years of service for your shaking needs.