Learn More
Selleck Chemical LLC BS-181 HCl 50mg 1397219-81-6
SDP
Supplier:  Selleck Chemical LLC S157250MG
Encompass_Preferred
BS-181 HCl is a highly selective CDK7 inhibitor with IC50 of 21 nM. It is more than 40-fold selective for CDK7 than CDK1, 2, 4, 5, 6, or 9. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-713-6