Learn More
Cayman Chemical BS-181 (hydrochloride), Purity: ≥98%, CAS: 1397219-81-6, 10 mg
SDP
Catalog No. 502966824
Encompass_Preferred
A selective Cdk7 inhibitor that blocks the activity of Cdk-activating kinase (IC50 = 21 nM); inhibits the growth of cancer cell lines in vitro and MCF-7 human xenografts in nude mice
Catalog No. 50-296-6824 Supplier Cayman Chemical Supplier No. 1799610 MG
A selective Cdk7 inhibitor that blocks the activity of Cdk-activating kinase (IC50 = 21 nM); inhibits the growth of cancer cell lines in vitro and MCF-7 human xenografts in nude mice