Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BS3 crosslinker | 82436-77-9 | MFCD01861948 | 99.8% | 528.47 | C16H20N2O14S2 | 100 MG
SDP

Catalog No. 5000201908
Encompass_Preferred

BS3 Crosslinker is a water-soluble, homobifunctional N-hydroxysuccinimide (NHS)-ester crosslinker used to covalently link primary amines in proteins and small molecules. It provides a non-cleavable eight-carbon spacer arm appropriate for antibody-drug conjugate synthesis, bioconjugation, and protein crosslinking workflows. The reagent offers high purity, documented solubility in common solvents, and manufacturer guidance for storage and handling to preserve stability.

  • Non-cleavable homobifunctional NHS-ester crosslinker.
  • Water soluble, suitable for aqueous bioconjugation.
  • Reacts specifically with primary amines to form stable amide bonds.
  • Eight-carbon spacer arm for controlled linker length.
  • High purity (99.84%) for reproducible results.
  • Soluble in DMSO and water; ultrasonic agitation recommended.
  • Storage recommendations included to preserve stability.

Catalog No. 50-002-01908 Supplier Medchemexpress LLC Supplier No. HY124329100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BS3 Crosslinker is a water-soluble, homobifunctional N-hydroxysuccinimide (NHS)-ester crosslinker used to covalently link primary amines in proteins and small molecules. It provides a non-cleavable eight-carbon spacer arm appropriate for antibody-drug conjugate synthesis, bioconjugation, and protein crosslinking workflows. The reagent offers high purity, documented solubility in common solvents, and manufacturer guidance for storage and handling to preserve stability.

  • Non-cleavable homobifunctional NHS-ester crosslinker.
  • Water soluble, suitable for aqueous bioconjugation.
  • Reacts specifically with primary amines to form stable amide bonds.
  • Eight-carbon spacer arm for controlled linker length.
  • High purity (99.84%) for reproducible results.
  • Soluble in DMSO and water; ultrasonic agitation recommended.
  • Storage recommendations included to preserve stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.