Learn More
Medchemexpress LLC BS3 crosslinker disodium | 127634-19-9 | 98.0% | 572.43 g/mol | C16H18N2Na2O14S2 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY124329A25MG
Encompass_Preferred
BS3 crosslinker disodium is a water-soluble, non-cleavable homobifunctional crosslinker with sulfo-NHS ester reactive groups and an 8-carbon spacer. It targets primary amines to form stable amide bonds, enabling efficient protein-protein crosslinking and antibody-drug conjugate synthesis in aqueous conditions. The reagent is provided as a sodium salt and is suitable for bioconjugation, analytical crosslinking, and ADC linker applications.
  • Water-soluble, compatible with aqueous reactions.
  • Reacts with primary amines to form stable amide bonds.
  • Non-cleavable linker for stable conjugates.
  • Sulfo-NHS ester groups enable efficient coupling at physiological pH.
  • High purity supports reproducible bioconjugation.
  • Soluble in DMSO and tested in common in vivo vehicle systems.
  • Stable when stored sealed at low temperatures away from moisture and light.
Catalog No. 50-002-02973