Learn More
Medchemexpress LLC BS3 crosslinker (disodium) | 127634-19-9 | MFCD00054982 | 98.0% | 572.43 | C16H18N2Na2O14S2 | 100 MG
SDP
Catalog No. 5000204207
Encompass_Preferred
BS3 crosslinker disodium is a water-soluble, non-cleavable homobifunctional N-hydroxysuccinimide (NHS) ester that selectively reacts with primary amines to form stable amide linkages. It is intended for research use in protein crosslinking and antibody-drug conjugation. Handle according to the SDS and protect from moisture and light during storage.
  • Water-soluble, homobifunctional NHS ester
  • Reacts with primary amines to form stable, non-reducible amide bonds
  • Suitable for protein crosslinking and antibody-drug conjugation workflows
  • High purity for reproducible experimental results
  • Available in multiple package sizes for varied experimental scales
  • Store at -20°C sealed and protected from moisture and light; in solvent, store at -80°C for long-term storage
Catalog No. 50-002-04207 Supplier Medchemexpress LLC Supplier No. HY124329A100MG
BS3 crosslinker disodium is a water-soluble, non-cleavable homobifunctional N-hydroxysuccinimide (NHS) ester that selectively reacts with primary amines to form stable amide linkages. It is intended for research use in protein crosslinking and antibody-drug conjugation. Handle according to the SDS and protect from moisture and light during storage.
  • Water-soluble, homobifunctional NHS ester
  • Reacts with primary amines to form stable, non-reducible amide bonds
  • Suitable for protein crosslinking and antibody-drug conjugation workflows
  • High purity for reproducible experimental results
  • Available in multiple package sizes for varied experimental scales
  • Store at -20°C sealed and protected from moisture and light; in solvent, store at -80°C for long-term storage