Learn More
Medchemexpress LLC BS3 crosslinker (disodium) | 127634-19-9 | not stated | 98.0% | 572.43 g/mol | C16H18N2Na2O14S2 | 50 MG
SDP
Catalog No. 5000204050
Encompass_Preferred
BS3 Crosslinker disodium is a water-soluble, non-cleavable homobifunctional amine crosslinker used for bioconjugation and antibody-drug conjugate synthesis. It contains sulfosuccinimidyl reactive esters that selectively react with primary amines to form stable amide bonds, enabling protein crosslinking and labeling under aqueous conditions. The reagent is supplied as a solid of high purity and requires sealed storage away from moisture and light; in solution, long-term storage at -80°C is recommended.
Catalog No. 50-002-04050 Supplier Medchemexpress LLC Supplier No. HY124329A50MG
BS3 Crosslinker disodium is a water-soluble, non-cleavable homobifunctional amine crosslinker used for bioconjugation and antibody-drug conjugate synthesis. It contains sulfosuccinimidyl reactive esters that selectively react with primary amines to form stable amide bonds, enabling protein crosslinking and labeling under aqueous conditions. The reagent is supplied as a solid of high purity and requires sealed storage away from moisture and light; in solution, long-term storage at -80°C is recommended.