Learn More
TARGETMOL CHEMICALS INC BS3 Crosslinker disodium 25MG
SDP
Catalog No. 502878255
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BS3 Crosslinker disodium, a non-cleavable ADC linker, is employed in the synthesis of antibody-drug conjugates (ADCs). Purity 97.94%
Catalog No. 50-287-8255 Supplier TARGETMOL CHEMICALS INC Supplier No. T14833L25MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. BS3 Crosslinker disodium, a non-cleavable ADC linker, is employed in the synthesis of antibody-drug conjugates (ADCs). Purity 97.94%