Learn More
Abcam BS3 crosslinker, primary amine reactive crosslinker, 50MG
Catalog No. 502204963
Encompass_Preferred
MW 572.4 Da, Purity >90%. Homobifunctional, membrane-impermeable primary amine reactive crosslinker. Crosslinks cell surface proteins. Facilitates structural characterization of proteins with mass spectrometry.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-4963 Supplier Abcam Supplier No. AB14561250MG
Add to Cart
Add to Cart
MW 572.4 Da, Purity >90%. Homobifunctional, membrane-impermeable primary amine reactive crosslinker. Crosslinks cell surface proteins. Facilitates structural characterization of proteins with mass spectrometry.
The product is subject to the following: Abcam Restricted Use Statement