Learn More
Bioworld BSA (1%) IN PBS, DNASE AND RNA
Catalog No. NC2193296
Encompass_Preferred
BSA (1%) in PBS, DNase and RNase Free0.79% Sodium Chloride, 0.02% Potassium Chloride, 0.06% Sodium Phosphate, 0.02% Potassium Phosphate, 1.0% BSA, DNAse/RNAse-Free
This item is not returnable. View return policy
Catalog No. NC2193296 Supplier Bioworld Supplier No. 401207061
Add to Cart
Add to Cart
BSA (1%) in PBS, DNase and RNase Free0.79% Sodium Chloride, 0.02% Potassium Chloride, 0.06% Sodium Phosphate, 0.02% Potassium Phosphate, 1.0% BSA, DNAse/RNAse-Free