Learn More
STEMCELL Technologies 10% Bovine Serum Albumin in Iscove's MDM, Size: 100 mL
SDP
Catalog No. NC9179618
Encompass
This product has been developed for use in applications where a culture medium of defined composition is required. It contains pre-screened batches of bovine serum albumin (BSA) that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations. It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions.Recommended for use with MethoCult™ H4100 or MethoCult™ M3134 for hematopoietic colony-forming unit (CFU) assays (at a final concentration of 1%).
Catalog No. NC9179618 Supplier STEMCELL Technologies Supplier No. 09300
This product has been developed for use in applications where a culture medium of defined composition is required. It contains pre-screened batches of bovine serum albumin (BSA) that have been selected to support the optimal growth of human hematopoietic progenitor cells in serum-free media formulations. It is also suitable for the culture of mouse hematopoietic progenitor cells in serum-free conditions.Recommended for use with MethoCult™ H4100 or MethoCult™ M3134 for hematopoietic colony-forming unit (CFU) assays (at a final concentration of 1%).