Learn More
Bioworld Bovine Serum Albumin 20%, 9048-46-8 , Sterile, 50ml
Supplier:  Bioworld 402200521
Encompass_Preferred
A general purpose purified albumin, suitable for Westerns, enzyme systems and as a protein supplement. Specifications: 20% BSA solution in 0.85% NaCl, Biotechnology Grade Bovine Serum Albumin, Sterile filtered, HS Code : 350290700, Molecular Weight : 66.43, pH : 7.0-7.4, Solubility : Soluble in water, Appearance Form : Liquid, Appearance Color : Clear/yellow. 50ml
Catalog No. 50-255-465