Learn More
Bioworld Bovine Serum Albumin (BSA), 20%, Sterile, 500 mL
Supplier:  Bioworld 402200522
Encompass_Preferred
A general purpose purified albumin, suitable for Westerns, enzyme systems and as a protein supplement 20% BSA solution in 0.85% NaClBiotechnology Grade Bovine Serum AlbuminSterile filteredHS Code : 350290700
Catalog No. 50-255-466