Learn More
Bioworld Bovine Serum Albumin (BSA), 20 mg/mL, 5 mL
Supplier:  Bioworld 402200561
Encompass_Preferred
- Biotechnology Grade Bovine Serum Albumin (BSA)A general purpose purified albumin, suitable for Westerns, enzyme systems and as a protein supplementAbs.@280nm (1:40, Water)- 0.25-0.35 DNase(P/F)- None Electrophoresis(P/F)- Pass Protease(P/F)- None RNase(P/F)- None Total Protein(mg/mL)- 18-22Appearance: Liquid
Catalog No. 50-255-467