Learn More
Cygnus Technologies BSA Antigen Concentrate, 1 mL
Catalog No. NC1782939
Encompass
For use in spike and recovery studies and as a positive control.
Catalog No. NC1782939 Supplier Cygnus Technologies Supplier No. F031G
For use in spike and recovery studies and as a positive control.