Learn More
Bioworld BSA-BLOCKING BUFFER IN PBS
Catalog No. 50255471
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. BSA-Blocking Buffer in PBS (10X) Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 3% BSA in PBS; Sodium Azide (0.02%).
Catalog No. 50-255-471 Supplier Bioworld Supplier No. 402200641
This item has a minimum order quantity of 2 per supplier requirements. BSA-Blocking Buffer in PBS (10X) Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 3% BSA in PBS; Sodium Azide (0.02%).