Learn More
Bioworld BSA-BLOCKING BUFFER IN PBS
Supplier:  Bioworld 402200642
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. BSA-Blocking Buffer in PBS (10X) Blot blocking buffer used to block non-specific binding site for antibodies on the membrane Technical Information: 3% BSA in PBS; Sodium Azide (0.02%).
Catalog No. 50-255-472