Learn More
Bioworld BSA-BLOCKING BUFFER IN TBS
Supplier:  Bioworld 402200682
Encompass_Preferred
BSA-Blocking Buffer in TBS (10X) Blot blocking buffer used to block non-specific binding site for antibodies on the membrane 1x Solution Contains: 3% BSA in TBS (pH 7.4); Sodium Azide (0.02%).
Catalog No. 50-255-474