Learn More
Bioworld BSA-BLOCKING BUFFER IN TBS
Catalog No. 50255473
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements. BSA-Blocking Buffer in TBS (10X) Blot blocking buffer used to block non-specific binding site for antibodies on the membrane 1x Solution Contains: 3% BSA in TBS (pH 7.4); Sodium Azide (0.02%).
Catalog No. 50-255-473 Supplier Bioworld Supplier No. 402200681
This item has a minimum order quantity of 2 per supplier requirements. BSA-Blocking Buffer in TBS (10X) Blot blocking buffer used to block non-specific binding site for antibodies on the membrane 1x Solution Contains: 3% BSA in TBS (pH 7.4); Sodium Azide (0.02%).