Learn More
Alkali Scientific BSA Blocking Buffer Solution in PBS with Azide, 200 Milliliters
SDP
Catalog No. 5000660401
Encompass_Preferred
BSA Blocking Buffer Solution in PBS with Azide is a protein blocking buffer used in molecular biology and biochemistry applications. This 200mL solution is ideal for blocking non-specific binding sites during immunoassays such as Western blotting, ELISA, and immunohistochemistry. The buffer contains Bovine Serum Albumin (BSA) to prevent nonspecific binding of antibodies or other proteins to the membrane, ensuring clearer and more reliable results. PBS provides a stable pH and isotonic conditions for the samples, while azide acts as a preservative to extend the shelf life of the solution. This product is essential in protein analysis and detection experiments.
Catalog No. 50-006-60401 Supplier Alkali Scientific Supplier No. ABL001CP
BSA Blocking Buffer Solution in PBS with Azide is a protein blocking buffer used in molecular biology and biochemistry applications. This 200mL solution is ideal for blocking non-specific binding sites during immunoassays such as Western blotting, ELISA, and immunohistochemistry. The buffer contains Bovine Serum Albumin (BSA) to prevent nonspecific binding of antibodies or other proteins to the membrane, ensuring clearer and more reliable results. PBS provides a stable pH and isotonic conditions for the samples, while azide acts as a preservative to extend the shelf life of the solution. This product is essential in protein analysis and detection experiments.