Learn More
Research Products International Corp BSA Blocking Buffer Solution in PBS with Azide, 200 Milliliters
SDP
Catalog No. 50995801
Encompass_Preferred
Formulation: 3% BSA in PBS plus 0.02% Azide
Catalog No. 50-995-801 Supplier Research Products International Corp Supplier No. B41600200.0
Formulation: 3% BSA in PBS plus 0.02% Azide