Learn More
Bioworld Bovine Serum Albumin, Fraction V - Standard Grade, 9048-46-8, 100g
Catalog No. 50253900
Encompass_Preferred
Protein with numerous biochemical applications including ELISAs, immunoblots, and immunohistochemistry. Heat shock isolation. Specifications: pH (5% water): 6.5-7.5, Heavy Metals (Pb): <0.001%, DNase: None detected, RNase: None detected, Protease: None detected. This product is sold for laboratory use only. Solubility : Soluble in water. Appearance Form : Solid. Appearance Color : Light brown. 100g
Catalog No. 50-253-900 Supplier Bioworld Supplier No. 220700083
Protein with numerous biochemical applications including ELISAs, immunoblots, and immunohistochemistry. Heat shock isolation. Specifications: pH (5% water): 6.5-7.5, Heavy Metals (Pb): <0.001%, DNase: None detected, RNase: None detected, Protease: None detected. This product is sold for laboratory use only. Solubility : Soluble in water. Appearance Form : Solid. Appearance Color : Light brown. 100g