Learn More
Bioworld Bovine Serum Albumin (BSA), Crystallized, 500 g
Supplier:  Bioworld 220700045
Encompass_Preferred
Bovine Serum Albumin (BSA) Technical Information:Biotechnology gradeMoisture: < 1.0%Heavy metals: <0.5%pH (10% water): 6.5 - 7.5
Catalog No. 50-253-893