Learn More
Bioworld Bovine Serum Albumin (BSA) Standard, 2 mg/mL, 10 mL
Catalog No. 50253896
Encompass_Preferred
Bovine Albumin StandardStandard used for determining protein concentration by CB protein assay. 2 mg/mL dissolved in 0.9% sodium chloride with 0.1% sodium azide as a preservative and sterile filtered.
Catalog No. 50-253-896 Supplier Bioworld Supplier No. 220700061
Bovine Albumin StandardStandard used for determining protein concentration by CB protein assay. 2 mg/mL dissolved in 0.9% sodium chloride with 0.1% sodium azide as a preservative and sterile filtered.