Learn More
Bioworld Bovine Serum Albumin, RNase Free, 9048-46-8, 1ml
Supplier:  Bioworld 402000641
Encompass_Preferred
Bovine serum albumin, BSA, is a single subunit protein with no enzymatic activity and does not interfere with any molecular biology reactions.A general purpose purified albumin, suitable for Westerns, enzyme systems and as a protein supplement. Supplied at 10 mg/mL in 0.85% sodium chloride. Contains 0.1% sodium azide as preservative. Aseptically filled. APPLICATIONS: Protease-sensitive Immunoassays such as RIA, EIA, Fluorescent and Chemiluminescent, Protein Standard, Diluent, Protein, Conjugate or Enzyme Stabilizer, Hybridization, Selected Cell Culture applications. FEATURES AND BENEFITS: High Purity, Low Interference, Low Background, Virtually No Detectable IgG, Endotoxin, Enzyme or Protease Activity, Superior Solubility/Filterability, Validated for Prion (BSE/TSE) Clearance, Endotoxin (LAL): ≤ 0.1EU/mg max.
Catalog No. 50-255-395