Learn More
Bioworld Bovine Serum Albumin (BSA), A1, 25 g
Supplier:  Bioworld 220700072
Encompass_Preferred
Bovine Serum Albumin (BSA), Alcohol Isolation Specifications:CAS 9048-46-8Cold Alcohol Isolation Biotechnology gradePurity (Electrophoresis): >99%Moisture <5.0%DNase/RNase: None DetectedAsh: 0.5% Heavy Metals: 0.005% Storage: 2-8°C
Catalog No. 50-253-898