Learn More
Bioworld Bovine Serum Albumin (BSA), 1 mg/mL, 50 mL
Supplier:  Bioworld 220700052
Encompass_Preferred
1mg/ml BSA (Bovine serum albumin) solution in 0.85% Sodium Chloride and 0.01% Sodium azide.
Catalog No. 50-253-895