Learn More
Cayman Chemical BSA Control for BSA-Fatty Acid Complexes (5 mM), 5 mL
SDP
Catalog No. 502988522
Encompass_Preferred
A control for 5 mM BSA-fatty acid complexes; can be used as a control for experiments using BSA-fatty acid complexes to deliver fatty acids to cells in culture
Catalog No. 50-298-8522 Supplier Cayman Chemical Supplier No. 295565 ML
A control for 5 mM BSA-fatty acid complexes; can be used as a control for experiments using BSA-fatty acid complexes to deliver fatty acids to cells in culture