Learn More
Cayman Chemical BSA Control for BSA-Fatty Acid Complexes (1 mM), 5 mL
SDP
Catalog No. 502910545
Encompass_Preferred
A control for 1 mM BSA-fatty acid complexes; can be used as a control for experiments using BSA-fatty acid complexes to deliver fatty acids to cells in culture
Catalog No. 50-291-0545 Supplier Cayman Chemical Supplier No. 349325 ML
A control for 1 mM BSA-fatty acid complexes; can be used as a control for experiments using BSA-fatty acid complexes to deliver fatty acids to cells in culture