Learn More
Alpha Diagnostic International Bovine Albumin (BSA) ELISA Kit, 96 tests, Quantitative
SDP
Supplier:  Alpha Diagnostic International 8100
Encompass
The antibodies used in this kit have been shown by immunoelectrophoresis and ELISA to react specifically with BSA, and to have essentially no reactivity with other bovine serum proteins.
Catalog No. NC0455989