Learn More
Gold Biotechnology Inc BSA Fraction V Protease Free
SDP
Catalog No. 5000562033
Encompass_Preferred
GoldBio Caprylate Stabilized BSA is a highly purified lyophilized albumin suitable for Westerns enzyme systems protein standards hybridizations and as a protease-sensitive immunoassays It is also perfectly suitable as a nutrient in cell culture media with low interference and backgrounds and is ideal for most standard molecular biology experiments Our Caprylate Stabilized BSA is protease and certified TSE free manufactured in the United States in a closed loop system from USDA-inspected healthy animals located in the US
Catalog No. 50-005-62033 Supplier Gold Biotechnology Inc Supplier No. A42010
GoldBio Caprylate Stabilized BSA is a highly purified lyophilized albumin suitable for Westerns enzyme systems protein standards hybridizations and as a protease-sensitive immunoassays It is also perfectly suitable as a nutrient in cell culture media with low interference and backgrounds and is ideal for most standard molecular biology experiments Our Caprylate Stabilized BSA is protease and certified TSE free manufactured in the United States in a closed loop system from USDA-inspected healthy animals located in the US