Learn More
Genscript Corporation BSA His Bovine
Catalog No. 5000057504
Encompass
Bovine serum albumin (BSA) is a soluble monomeric protein It is widely as a supplement in biochemical and tissue culture media promoting cell growth and survival BSA stabilizes extracellular fluid volume and functions as a carrier for small molecules such as steroids fatty acids and thyroid hormones It is also used in drug development protein purification and food processing
Catalog No. 50-000-57504 Supplier Genscript Corporation Supplier No. Z03737100
Bovine serum albumin (BSA) is a soluble monomeric protein It is widely as a supplement in biochemical and tissue culture media promoting cell growth and survival BSA stabilizes extracellular fluid volume and functions as a carrier for small molecules such as steroids fatty acids and thyroid hormones It is also used in drug development protein purification and food processing