Learn More
Cayman Chemical BSA-Linoleate ω-6 Polyunsaturated Fatty Acid Complex (1 mM), 5 mL
SDP
Catalog No. 502919130
Encompass_Preferred
A BSA complex with linoleic acid; can be used as a source of essential nutrients to support cell growth in serum-free or reduced-serum media; can also be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring lipid metabolism and inflammatory signaling pathways
Catalog No. 50-291-9130 Supplier Cayman Chemical Supplier No. 386495 ML
A BSA complex with linoleic acid; can be used as a source of essential nutrients to support cell growth in serum-free or reduced-serum media; can also be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring lipid metabolism and inflammatory signaling pathways