Learn More
New England Biolabs, Inc. BSA-Molecular Biology Grade – 12 mg
SDP
Supplier:  New England Biolabs, Inc. B9000S
Encompass
Bovine Serum Albumin (BSA) is supplied with some products to prevent adhesion of the enzyme to reaction tubes and pipette surfaces. BSA also stabilizes some proteins during incubation.
Catalog No. NC0506695