Learn More
Enzo Life Sciences BSA (nitrated) (100 µg)
SDP
Supplier:  Enzo Life Sciences BMLSW1250100
Encompass_Preferred
Nitrated bovine serum albumin for use as a positive control for antibodies to nitrotyrosine. Nitrated bovine serum albumin for use as a positive control for antibodies to nitrotyrosine. Alternative name: Bovine serum albumin. Formulation: Lyophilized. Source: Native. Bovine albumin. Long Term Storage: -20°C. Applications: WB .
Catalog No. 50-201-2200