Learn More
Cayman Chemical BSA-Oleate Monounsaturated Fatty Acid Complex (5 mM), 5 mL
SDP
Catalog No. 502988892
Encompass_Preferred
A BSA complex with oleic acid; has been used for efficient fatty acid delivery to cells in culture for the purpose of monitoring lipid metabolism and inflammatory signaling pathways; has been used in the study of lipid accumulation in hepatic steatosis
Catalog No. 50-298-8892 Supplier Cayman Chemical Supplier No. 295575 ML
A BSA complex with oleic acid; has been used for efficient fatty acid delivery to cells in culture for the purpose of monitoring lipid metabolism and inflammatory signaling pathways; has been used in the study of lipid accumulation in hepatic steatosis