Learn More
Cayman Chemical BSA-Palmitate Saturated Fatty Acid Complex (5 mM), 5 mL
SDP
Catalog No. 502988818
Encompass_Preferred
A BSA complex with palmitic acid; can be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring fatty acid oxidation, lipid metabolism, and inflammatory signaling pathways; has been used to study the effect of long-chain fatty acid uptake on gene and protein expression, lipid droplet formation, hepatocyte lipid accumulation, and oxidative stress
Catalog No. 50-298-8818 Supplier Cayman Chemical Supplier No. 295585 ML
A BSA complex with palmitic acid; can be used for efficient fatty acid delivery to cells in culture for the purpose of monitoring fatty acid oxidation, lipid metabolism, and inflammatory signaling pathways; has been used to study the effect of long-chain fatty acid uptake on gene and protein expression, lipid droplet formation, hepatocyte lipid accumulation, and oxidative stress