Learn More
Enzo Life Sciences BSA solution (10%), (50 ml) (50 ml)
SDP
Supplier:  Enzo Life Sciences ADI801928
Encompass_Preferred
This BSA Solution (10%) is specially designed to create the Blocking Buffer and Assay Buffer in IMMUNOSET products. Each bottle is sufficient for use with 5x96 wells microtiter plates if following the instructions in our IMMUNOSET protocol. The BSA Solution may be used for any other assay development needs. Additional assay development reagents are available as companion products to our IMMUNOSET products. Shipping: Blue Ice Not Frozen. Long Term Storage: +4°C
Catalog No. 50-658-089