Learn More
MOLECULAR DEPOT LLC BSA Standard 200 mg/mL - 5 mL
Supplier:  MOLECULAR DEPOT LLC B2010660
Encompass_Preferred
BSA Protein Standard (200 mg/mL) is highly concentrated traceable standard solution of BSA at 200 mg/mL This product can be used in protein quantification This product is for research use only and is not for diagnostic applications
Catalog No. 50-000-13324