Learn More
Bioworld Bovine Serum Albumin (BSA) Standards Kit, 1 Kit
Catalog No. 50198429
Encompass_Preferred
Contains 8 x 1 ml BSA standards ranging from 1 mg/ml to 8 mg/ml concentration
Catalog No. 50-198-429 Supplier Bioworld Supplier No. 208300031
Contains 8 x 1 ml BSA standards ranging from 1 mg/ml to 8 mg/ml concentration