Learn More
Research Products International Corp BSA-Tween 20 Blocking Solution in TBS, 1 Liter
SDP
Catalog No. 50488568
Encompass_Preferred
Contains 3% BSA and 0.05% Tween 20 in TBS buffer. Optimized for use in western blotting to minimize non-specific binding of antibodies.
Catalog No. 50-488-568 Supplier Research Products International Corp Supplier No. B820001000.0
Contains 3% BSA and 0.05% Tween 20 in TBS buffer. Optimized for use in western blotting to minimize non-specific binding of antibodies.