Learn More
Research Products International Corp BSA-Tween 20 Blocking Solution in TBS, 500 Milliliters
SDP
Catalog No. 50488569
Encompass_Preferred
Contains 3% BSA and 0.05% Tween 20 in TBS buffer. Optimized for use in western blotting to minimize non-specific binding of antibodies.
Catalog No. 50-488-569 Supplier Research Products International Corp Supplier No. B82000500.0
Contains 3% BSA and 0.05% Tween 20 in TBS buffer. Optimized for use in western blotting to minimize non-specific binding of antibodies.