Learn More
New England Biolabs, Inc. BsaAI – 500 units
SDP
Catalog No. 50812194
Encompass
A restriction endonuclease that recognizes the sequence YAC_^GTR.
Catalog No. 50-812-194 Supplier New England Biolabs, Inc. Supplier No. R0531S
A restriction endonuclease that recognizes the sequence YAC_^GTR.