Learn More
New England Biolabs, Inc. BsaBI – 2000 units
SDP
Catalog No. 50812207
Encompass
A restriction endonuclease that recognizes the sequence GATNN_^NNATC.
Catalog No. 50-812-207 Supplier New England Biolabs, Inc. Supplier No. R0537S
Add to Cart
Add to Cart
A restriction endonuclease that recognizes the sequence GATNN_^NNATC.