Learn More
New England Biolabs, Inc. BsaHI – 2000 units
SDP
Catalog No. 50812237
Encompass
A restriction endonuclease that recognizes the sequence GR^CG_YC.
Catalog No. 50-812-237 Supplier New England Biolabs, Inc. Supplier No. R0556S
A restriction endonuclease that recognizes the sequence GR^CG_YC.